9,9'-(5-Bromo-1,3-phenylene)bis(9H-carbazole) ≥97.0% (by HPLC, total nitrogen)

Leverandør: TCI

Synonymer: 1-Bromo-3,5-bis(carbazol-9-yl)benzene

B5502-1G B5502-5G
TCIAB5502-1GEA 1175 NOK
TCIAB5502-1G TCIAB5502-5G
9,9'-(5-Bromo-1,3-phenylene)bis(9H-carbazole) ≥97.0% (by HPLC, total nitrogen)
9,9'-(5-Bromo-1,3-phenylene)bis(9H-carbazole)
Formel: C₃₀H₁₉BrN₂
MW: 487,40 g/mol
MDL nummer: MFCD28100481
CAS-nummer: 750573-24-1

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (with Total Nitrogen) min. 97.0 %
Melting point 224.0 to 228.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR