1-Benzoylpiperidine ≥98.0% (by GC)
Leverandør: TCI
Synonymer:
Phenyl(piperidin-1-yl)methanone
B5512-1G
B5512-5G
TCIAB5512-1GEA
325
NOK
TCIAB5512-1G
TCIAB5512-5G
1-Benzoylpiperidine ≥98.0% (by GC)
1-Benzoylpiperidine
Formel:
C₁₂H₁₅NO MW: 189,26 g/mol Kokepunkt: 321°C |
MDL nummer:
MFCD00023702 CAS-nummer: 776-75-0 EINECS: 212-280-3 |
Spesifikasjon Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 48.0 to 52.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...