1,2-Bis[(3,6-dibromo-9H-carbazol-9-yl)methyl]benzene ≥98.0% (by HPLC)

Leverandør: TCI

Synonymer: 9,9'-[1,2-Phenylenebis(methylene)]bis(3,6-dibromo-9H-carbazole)

B5605-1G B5605-5G
TCIAB5605-1GEA 675 NOK
TCIAB5605-1G TCIAB5605-5G
1,2-Bis[(3,6-dibromo-9H-carbazol-9-yl)methyl]benzene ≥98.0% (by HPLC)
1,2-Bis[(3,6-dibromo-9H-carbazol-9-yl)methyl]benzene
Formel: C₃₂H₂₀Br₄N₂
MW: 752,14 g/mol
Smeltepunkt: 319°C
CAS-nummer: 222166-46-3

Bestill nå

Spesifikasjon Testresultater

Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 3.30 to 4.10 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR