9,9-Bis(4-methoxyphenyl)-9H-fluorene ≥98.0% (by GC)
Leverandør: TCI
B5617-1G
B5617-5G
TCIAB5617-1GEA
1875
NOK
TCIAB5617-1G
TCIAB5617-5G
9,9-Bis(4-methoxyphenyl)-9H-fluorene ≥98.0% (by GC)
9,9-Bis(4-methoxyphenyl)-9H-fluorene
Formel:
C₂₇H₂₂O₂ MW: 378,47 g/mol |
CAS-nummer:
117766-40-2 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 126.0 to 130.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...