3,3-Bis(bromomethyl)oxetane ≥95.0% (by GC)
Leverandør: TCI
Synonymer:
2,2-Bis(bromomethyl)-1,3-epoxypropane
B5641-1G
B5641-5G
TCIAB5641-1GEA
515
NOK
TCIAB5641-1G
TCIAB5641-5G
3,3-Bis(bromomethyl)oxetane ≥95.0% (by GC)
3,3-Bis(bromomethyl)oxetane
Formel:
C₅H₈Br₂O MW: 243,93 g/mol Kokepunkt: 125°C Smeltepunkt: 25°C |
MDL nummer:
MFCD00269694 CAS-nummer: 2402-83-7 |
Spesifikasjon Testresultater
Purity (GC) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...