1,11-Bis(maleimido)-3,6,9-trioxaundecane ≥98.0% (by HPLC)

Leverandør: TCI

Synonymer: 1,11-Bis(maleimido)triethylene Glycol, BM-PEG3

B5668-100MG B5668-25MG
TCIAB5668-100MGEA 2225 NOK
TCIAB5668-100MG TCIAB5668-25MG
1,11-Bis(maleimido)-3,6,9-trioxaundecane ≥98.0% (by HPLC)
1,11-Bis(maleimido)-3,6,9-trioxaundecane
Formel: C₁₆H₂₀N₂O₇
MW: 352,34 g/mol
Lagringstemperatur: Fryseskap
MDL nummer: MFCD01863352
CAS-nummer: 86099-06-1

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 7.40 to 8.40 %
Melting point 63.0 to 68.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR