2'-Bromo-5'-phenyl-1,1':3',1''-terphenyl ≥98.0% (by GC)
Leverandør: TCI
Synonymer:
2-Bromo-1,3,5-triphenylbenzene, 2'-Bromo-5'-phenyl-m-terphenyl
B5674-200MG
B5674-1G
TCIAB5674-200MGEA
1325
NOK
TCIAB5674-200MG
TCIAB5674-1G
2'-Bromo-5'-phenyl-1,1':3',1''-terphenyl ≥98.0% (by GC)
2'-Bromo-5'-phenyl-1,1':3',1''-terphenyl
Formel:
C₂₄H₁₇Br MW: 385,30 g/mol |
MDL nummer:
MFCD00092320 CAS-nummer: 10368-73-7 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 128.0 to 132.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...