(R)-5-Bromo-3-[1-(2,6-dichloro-3-fluorophenyl)ethoxy]pyridin-2-amine ≥98.0% (by HPLC)

Leverandør: TCI
Warning

Synonymer: (R)-5-Bromo-3-(1-(2,6-dichloro-3-fluorophenyl)ethoxy)pyridin-2-amine

B5682-1G B5682-5G
TCIAB5682-1GEA 1100 NOK
TCIAB5682-1G TCIAB5682-5G
(R)-5-Bromo-3-[1-(2,6-dichloro-3-fluorophenyl)ethoxy]pyridin-2-amine ≥98.0% (by HPLC)
(R)-5-Bromo-3-[1-(2,6-dichloro-3-fluorophenyl)ethoxy]pyridin-2-amine
Formel: C₁₃H₁₀BrCl₂FN₂O
MW: 380,04 g/mol
MDL nummer: MFCD18207061
CAS-nummer: 877399-00-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=055157ffa010da4964357ba906cb35ba40808a46&vid=055157ffa010da4964357ba906cb35ba40808a46&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Specific rotation (value) -64.0 to -70.0 deg (C=1, CHCL3)
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 108.0 to 113.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR