5,5''-Bis(trimethylstannyl)-2,2':5',2''-terthiophene ≥98.0% (by HPLC)
Leverandør: TCI
|
Danger
|
B5690-200MG
B5690-1G
TCIAB5690-200MGEA
1550
NOK
TCIAB5690-200MG
TCIAB5690-1G
5,5''-Bis(trimethylstannyl)-2,2':5',2''-terthiophene ≥98.0% (by HPLC)
5,5''-Bis(trimethylstannyl)-2,2':5',2''-terthiophene
Formel:
C₁₈H₂₄S₃Sn₂ MW: 573,99 g/mol Lagringstemperatur: Kjøleskap |
MDL nummer:
MFCD30489177 CAS-nummer: 178931-63-0 FN: 3146 ADR: 6.1,I |
Spesifikasjon Testresultater
Appearance | White to Yellow to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Melting point | 120.0 to 124.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...