1-Bromo-4-klor-2-nitrobensen ≥98.0% (by GC)

Leverandør: TCI
Warning

B5696-5G B5696-25G
TCIAB5696-5GEA 380 NOK
TCIAB5696-5G TCIAB5696-25G
1-Bromo-4-klor-2-nitrobensen ≥98.0% (by GC)
1-Bromo-4-klor-2-nitrobensen
Formel: C₆H₃BrClNO₂
MW: 236,45 g/mol
MDL nummer: MFCD00024320
CAS-nummer: 41513-04-6
EINECS: 255-421-4

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 67.0 to 71.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR