1,4-Bis[4-(N,N-diphenylamino)styryl]benzene ≥98.0% (by HPLC, total nitrogen)
Leverandør: TCI
Synonymer:
DSA-Ph, 1,4-Bis(4-diphenylaminostyryl)benzene
B5820-200MG
B5820-1G
TCIAB5820-200MGEA
1500
NOK
TCIAB5820-200MG
TCIAB5820-1G
1,4-Bis[4-(N,N-diphenylamino)styryl]benzene ≥98.0% (by HPLC, total nitrogen)
1,4-Bis[4-(N,N-diphenylamino)styryl]benzene
Formel:
C₄₆H₃₆N₂ MW: 616,81 g/mol Smeltepunkt: 172°C |
MDL nummer:
MFCD12022510 CAS-nummer: 55035-42-2 |
Spesifikasjon Testresultater
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 228.0 to 232.0 deg-C |
Lambda max. | 408.0 to 414.0 nm (toluene) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...