SB239063 ≥98.0% (by HPLC)
Leverandør: TCI
Synonymer:
4-[4-(4-fluorophenyl)-5-(2-methoxypyrimidin-4-yl)imidazol-1-yl]cyclohexan-1-ol, trans-4-[4-(4-fluorophenyl)-5-(2-methoxy-4-pyrimidinyl)-1H-imidazol-1-yl]-cyclohexanol, Rel-(1r,4r)-4-(4-(4-fluorophenyl)-5-(2-methoxypyrimidin-4-yl)-1H-imidazol-1-yl)cyclohexan-1-ol, 4-(4-(4-fluorophenyl)-5-(2-methoxypyrimidin-4-yl)-1H-imidazol-1-yl)cyclohexanol
B5898-25MG
B5898-5MG
TCIAB5898-25MGEA
5800
NOK
TCIAB5898-25MG
TCIAB5898-5MG
SB239063 ≥98.0% (by HPLC)
SB239063
Formel:
C₂₀H₂₁FN₄O₂ MW: 368,4 g/mol Lagringstemperatur: Fryseskap |
MDL nummer:
MFCD03792786 CAS-nummer: 193551-21-2 |
Spesifikasjon Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...