Bis(2-chloroethyl)phosphoramidic Dichloride ≥98.0% (by GC)

Leverandør: TCI
Danger

B5908-1G B5908-5G
TCIAB5908-1GEA 1500 NOK
TCIAB5908-1G TCIAB5908-5G
Bis(2-chloroethyl)phosphoramidic Dichloride ≥98.0% (by GC)
Bis(2-chloroethyl)phosphoramidic Dichloride
Formel: C₄H₈Cl₄NOP
MW: 258,89 g/mol
Kokepunkt: 125°C
MDL nummer: MFCD00191796
CAS-nummer: 127-88-8
FN: 3261
ADR: 8,II

Bestill nå

Spesifikasjon Testresultater

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 97.0 %
Melting point 52.0 to 56.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR