O-(tert-Butyldiphenylsilyl)hydroxylamine ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: (tert-Butyldiphenylsilyloxy)amine

B5999-200MG B5999-1G
TCIAB5999-200MGEA 475 NOK
TCIAB5999-200MG TCIAB5999-1G
O-(tert-Butyldiphenylsilyl)hydroxylamine ≥98.0% (by GC)
O-(tert-Butyldiphenylsilyl)hydroxylamine
Formel: C₁₆H₂₁NOSi
MW: 271,44 g/mol
MDL nummer: MFCD06809831
CAS-nummer: 103587-51-5

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 87.0 to 91.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR