Trihexyl O-Butyrylcitrate ≥92.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
Butyryl trihexyl Citrate, Butyrylcitric acid trihexyl ester, Trihexyl 2-(Butyryloxy)propane-1,2,3-tricarboxylate, 2-(Butyryloxy)propane-1,2,3-tricarboxylic acid trihexyl ester
B6040-25G
B6040-500G
TCIAB6040-25GEA
220
NOK
TCIAB6040-25G
TCIAB6040-500G
Trihexyl O-Butyrylcitrate ≥92.0% (by GC)
Trihexyl O-Butyrylcitrate
Formel:
C₂₈H₅₀O₈ MW: 514,70 g/mol Flash punkt: 234°C |
MDL nummer:
MFCD00192535 CAS-nummer: 82469-79-2 EINECS: 413-890-4 FN: 3082 ADR: 9,III |
Spesifikasjon Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 92.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...