SB-505124 ≥98.0% (by HPLC)

Leverandør: TCI

Synonymer: 2-[5-(Benzo[d][1,3]dioxol-5-yl)-2-(tert-butyl)-1H-imidazol-4-yl]-6-methylpyridine, 2-(4-(Benzo[d][1,3]dioxol-5-yl)-2-(tert-butyl)-1H-imidazol-5-yl)-6-methylpyridine, 2-[4-(1,3-Benzodioxol-5-yl)-2-(tert-butyl)-1H-imidazol-5-yl]-6-methylpyridine

B6056-25MG B6056-5MG
TCIAB6056-25MGEA 4575 NOK
TCIAB6056-25MG TCIAB6056-5MG
SB-505124 ≥98.0% (by HPLC)
SB-505124
Formel: C₂₀H₂₁N₃O₂
MW: 335,4 g/mol
MDL nummer: MFCD20926329
CAS-nummer: 694433-59-5

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Light orange powder to crystaline
Purity (HPLC) min. 98.0 area%
Melting point gt. 166.0 to 170.0 deg-C
Elemental analysis (Nitrogen) 11.90 to 13.20 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR