BX-471 ≥98.0% (by HPLC)
Leverandør: TCI
Synonymer:
(R)-1-[5-Chloro-2-[2-[4-(4-fluorobenzyl)-2-methylpiperazin-1-yl]-2-oxoethoxy]phenyl]urea, BX471, (R)-1-(5-Chloro-2-(2-(4-(4-fluorobenzyl)-2-methylpiperazin-1-yl)-2-oxoethoxy)phenyl)urea
B6060-25MG
B6060-5MG
TCIAB6060-25MGEA
6400
NOK
TCIAB6060-25MG
TCIAB6060-5MG
BX-471 ≥98.0% (by HPLC)
BX-471
Formel:
C₂₁H₂₄ClFN₄O₃ MW: 434,89 g/mol Lagringstemperatur: Kjøleskap |
MDL nummer:
MFCD09859709 CAS-nummer: 217645-70-0 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Elemental analysis (Nitrogen) | 11.60 to 13.50 % |
Water (Vaporization methoed) | max. 8.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...