Benzyl isovalerate ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
Benzyl 3-methylbutanoate
B6099-100G
B6099-25G
TCIAB6099-100GEA
890
NOK
TCIAB6099-100G
TCIAB6099-25G
Benzyl isovalerate ≥98.0% (by GC)
Benzyl isovalerate
Formel:
C₁₂H₁₆O₂ MW: 192,26 g/mol Kokepunkt: 245°C |
MDL nummer:
MFCD00048348 CAS-nummer: 103-38-8 EINECS: 203-106-7 |
Spesifikasjon Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...