Dibutyl carbonate ≥98.0% (by GC)
Leverandør: TCI
Synonymer:
Carbonic acid dibutyl ester
C0040-25G
C0040-5G
TCIAC0040-25GEA
1975
NOK
TCIAC0040-25G
TCIAC0040-5G
Dibutyl carbonate ≥98.0% (by GC)
Dibutyl carbonate
Formel:
C₉H₁₈O₃ MW: 174,24 g/mol Kokepunkt: 207°C |
MDL nummer:
MFCD00043938 CAS-nummer: 542-52-9 EINECS: 208-816-0 Merck Indeks: 14,01558 |
Spesifikasjon Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...