N-Chloroacetyl-L-leucine ≥99.0% (by titrimetric analysis)
Leverandør: TCI
|
Warning
|
C0101-1G
C0101-25G
TCIAC0101-1GEA
300
NOK
TCIAC0101-1G
TCIAC0101-25G
N-Chloroacetyl-L-leucine ≥99.0% (by titrimetric analysis)
N-Chloroacetyl-L-leucine
Formel:
C₈H₁₄ClNO₃ MW: 207,65 g/mol |
MDL nummer:
MFCD00018912 CAS-nummer: 688-12-0 EINECS: 211-700-2 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | min. 99.0 % |
Melting point | 132.0 to 135.0 deg-C |
Specific rotation [a]20/D | -14.5 to -16.5 deg (C=2, EtOH) |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...