Carboxymethoxylamine hemihydrochloride ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Warning

Synonymer: Aminooxyacetic acid hemihydrochloride, O-(Carboxymethyl)hydroxylamine Hemihydrochloride, O-(Carboxymethyl)hydroxylamine hemihydrochloride, (Aminooxy)acetic Acid Hemihydrochloride

C0205-25G C0205-500G
TCIAC0205-25GEA 1925 NOK
TCIAC0205-25G TCIAC0205-500G
Carboxymethoxylamine hemihydrochloride ≥98.0% (by titrimetric analysis)
Carboxymethoxylamine hemihydrochloride
Formel: C₄H₁₁ClN₂O₆
MW: 218,59 g/mol
MDL nummer: MFCD00012955
CAS-nummer: 2921-14-4
EINECS: 220-862-3

Bestill nå

Spesifikasjon Testresultater

Appearance White powder to crystal
Purity (Neutralization titration) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Solubility in Water very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR