N-(2-Cyanoethyl)-N-ethylaniline ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: 3-(Ethyl(phenyl)amino)propanenitrile, 3-(N-Ethyl-N-phenylamino)propionitrile

C0451-25G C0451-500G
TCIAC0451-25GEA 220 NOK
TCIAC0451-25G TCIAC0451-500G
N-(2-Cyanoethyl)-N-ethylaniline ≥98.0% (by GC, titration analysis)
N-(2-Cyanoethyl)-N-ethylaniline
Formel: C₁₁H₁₄N₂
MW: 174,25 g/mol
Kokepunkt: 176°C
MDL nummer: MFCD00019858
CAS-nummer: 148-87-8
EINECS: 205-728-4

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Yellow to Green clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR