Carboxymethyl cellulose sodium salt n=∼1,050

Leverandør: TCI

Synonymer: Carboxymethyl cellulose, sodium salt, Cellulose gum, Cellulose, carboxymethyl ether, sodium salt, Sodium carboxymethyl cellulose (Viscosity 800mpa.s), Sodium carboxymethyl cellulose, CMC

C0603-25G C0603-500G
TCIAC0603-25GEA 230 NOK
TCIAC0603-25G TCIAC0603-500G
Carboxymethyl cellulose sodium salt n=∼1,050
Carboxymethyl cellulose sodium salt
Formel: [C₆H₇O₂(OH)x(OCH₂COONa)y]n
MDL nummer: MFCD00081472
CAS-nummer: 9004-32-4
Merck Indeks: 14,01829

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Content (Na,Drying substance) 6.0 to 8.5 %
Etherification value (as Drying substance) 0.5 to 0.8
Drying loss max. 10.0 %
Viscosity 900 to 1400 mPa-s (1 %, H2O, 25 deg-C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR