Vinyl chloroacetate ≥99.0% (by GC) stabilised

Leverandør: TCI
Danger

Synonymer: Chloroacetic Acid Vinyl Ester

Stabilisering: Stabilised with 4-methoxyphenol (MEHQ)

C0627-25G C0627-500G
TCIAC0627-25GEA 300 NOK
TCIAC0627-25G TCIAC0627-500G
Vinyl chloroacetate ≥99.0% (by GC) stabilised
Vinyl chloroacetate
Formel: C₄H₅ClO₂
MW: 120,53 g/mol
Kokepunkt: 136°C
Flash punkt: 51°C
MDL nummer: MFCD00045275
CAS-nummer: 2549-51-1
EINECS: 219-834-3
FN: 2589
ADR: 6.1,II

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR