Cyclohexyl vinyl ether ≥98.0% (by GC) stabilised
Leverandør: TCI
|
Warning
|
Stabilisering:
Stabilised with potassium hydroxide
C0885-25ML
C0885-500ML
TCIAC0885-25MLEA
270
NOK
TCIAC0885-25ML
TCIAC0885-500ML
Cyclohexyl vinyl ether ≥98.0% (by GC) stabilised
Cyclohexyl vinyl ether
Formel:
C₈H₁₄O MW: 126,20 g/mol Kokepunkt: 152°C Flash punkt: 35°C |
MDL nummer:
MFCD00046358 CAS-nummer: 2182-55-0 EINECS: 218-561-7 FN: 3271 ADR: 3,III |
Spesifikasjon Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...