5-(4-Chlorobutyl)hydantoin ≥97.0% (by titrimetric analysis)

Leverandør: TCI

C0902-25G C0902-5G
TCIAC0902-25GEA 2925 NOK
TCIAC0902-25G TCIAC0902-5G
5-(4-Chlorobutyl)hydantoin ≥97.0% (by titrimetric analysis)
5-(4-Chlorobutyl)hydantoin
Formel: C₇H₁₁ClN₂O₂
MDL nummer: MFCD00059722
CAS-nummer: 40126-55-4

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (Argentmetric Titration) min. 97.0 %
Melting point 128.0 to 132.0 deg-C
Solubility in Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR