Methyl-3-(4-methoxycarbonylphenyl)propionate ≥97.0% (by titrimetric analysis)
Leverandør: TCI
Synonymer:
3-(4-Carboxyphenyl)propionic Acid Dimethyl Ester, 3-(4-Methoxycarbonylphenyl)propionic Acid Methyl Ester, Methyl 4-(3-methoxy-3-oxopropyl)benzoate
C1029-25G
C1029-5G
TCIAC1029-25GEA
5350
NOK
TCIAC1029-25G
TCIAC1029-5G
Methyl-3-(4-methoxycarbonylphenyl)propionate ≥97.0% (by titrimetric analysis)
Methyl-3-(4-methoxycarbonylphenyl)propionate
Formel:
C₁₂H₁₄O₄ MW: 222,24 g/mol Lagringstemperatur: Kjøleskap |
MDL nummer:
MFCD00059305 CAS-nummer: 40912-11-6 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (Ester Value) | min. 97.0 % |
Melting point | 33.0 to 37.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...