Carbazole-9-carbonyl chloride ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: 9-(Chlorocarbonyl)carbazole

C1031-1G C1031-25G C1031-5G
TCIAC1031-1GEA 1250 NOK
TCIAC1031-1G TCIAC1031-25G TCIAC1031-5G
Carbazole-9-carbonyl chloride ≥98.0% (by GC, titration analysis)
Carbazole-9-carbonyl chloride
Formel: C₁₃H₈ClNO
MW: 229,66 g/mol
MDL nummer: MFCD00059689
CAS-nummer: 73500-82-0

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Melting point 103.0 to 106.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR