Carbimazole ≥95.0% (by HPLC)

Leverandør: TCI

Synonymer: 1-Ethoxycarbonyl-3-methyl-2-thioxo-4-imidazoline, Ethyl 3-methyl-2-thioxo-2,3-dihydro-1H-imidazole-1-carboxylate

C1080-25G C1080-5G C1080-1G
TCIAC1080-25GEA 3250 NOK
TCIAC1080-25G TCIAC1080-5G TCIAC1080-1G
Carbimazole ≥95.0% (by HPLC)
Carbimazole
Formel: C₇H₁₀N₂O₂S
MW: 186,23 g/mol
MDL nummer: MFCD00027421
CAS-nummer: 22232-54-8
EINECS: 244-854-4
Merck Indeks: 14,01798

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 95.0 area%
Melting point 122.0 to 125.0 deg-C
Thiamazole max. 0.5 w/w%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR