Cyanoacetylurea ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Danger

Synonymer: N-Carbamoyl-2-cyanoacetamide, Acetamide, N-(aminocarbonyl)-2-cyano-

C1101-25G C1101-250G
TCIAC1101-25GEA 260 NOK
TCIAC1101-25G TCIAC1101-250G
Cyanoacetylurea ≥98.0% (by titrimetric analysis)
Cyanoacetylurea
Formel: C₄H₅N₃O₂
MW: 127,10 g/mol
MDL nummer: MFCD00007947
CAS-nummer: 1448-98-2
EINECS: 215-906-3
FN: 3439
ADR: 6.1,III

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (Neutralization titration) min. 98.0 %
Solubility in hot Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR