Isopropyl crotonate ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: Crotonic Acid Isopropyl Ester, Isopropyl Crotonate

C1183-25ML C1183-500ML
TCIAC1183-25MLEA 715 NOK
TCIAC1183-25ML TCIAC1183-500ML
Isopropyl crotonate ≥98.0% (by GC)
Isopropyl crotonate
Formel: C₇H₁₂O₂
MW: 128,17 g/mol
Kokepunkt: 147°C
MDL nummer: MFCD00059053
CAS-nummer: 6284-46-4
FN: 3272
ADR: 3,III

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR