4-Chloro-4'-hydroxybiphenyl ≥98.0% (by titrimetric analysis)
Leverandør: TCI
|
Warning
|
Synonymer:
4'-Chloro-[1,1'-biphenyl]-4-ol, 4'-Chlorobiphenyl-4-ol
C1239-1G
C1239-5G
TCIAC1239-1GEA
1150
NOK
TCIAC1239-1G
TCIAC1239-5G
4-Chloro-4'-hydroxybiphenyl ≥98.0% (by titrimetric analysis)
4-Chloro-4'-hydroxybiphenyl
Formel:
C₁₂H₉ClO MW: 204,65 g/mol |
MDL nummer:
MFCD00143298 CAS-nummer: 28034-99-3 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 145.0 to 148.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...