N-Carbobenzoxy-O-tert-butyl-L-serine ≥98.0% (by titrimetric analysis)

Leverandør: TCI

Synonymer: Z-Ser(tBu)-OH, N-Cbz-O-tert-butyl-L-serine, (S)-2-(((Benzyloxy)carbonyl)amino)-3-(tert-butoxy)propanoic acid, Cbz-Ser-OtBu

C1381-1G C1381-5G
TCIAC1381-1GEA 285 NOK
TCIAC1381-1G TCIAC1381-5G
N-Carbobenzoxy-O-tert-butyl-L-serine ≥98.0% (by titrimetric analysis)
N-Carbobenzoxy-O-tert-butyl-L-serine
Formel: C₁₅H₂₁NO₅
MW: 295,34 g/mol
MDL nummer: MFCD00038275
CAS-nummer: 1676-75-1
EINECS: 216-827-7

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 85.0 to 89.0 deg-C
Specific rotation [a]20/D +19.0 to +23.0 deg (C=2, EtOH)
Solubility in EtOH almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR