Ethyl (R)-(-)-4-cyano-3-hydroxybutyate ≥97.0% (by GC)

Leverandør: TCI
Warning

Synonymer: (R)-4-Cyano-3-hydroxybutyric acid ethyl ester, BUTANOIC ACID, 4-CYANO-3-HYDROXY-, ETHYL ESTER, (3R)-, Ethyl (R)-4-cyano-3-hydroxybutanoate, (R)-ethyl 4-cyano-3-hydroxybutanoate

C1474-1G C1474-25G C1474-5G
TCIAC1474-1GEA 220 NOK
TCIAC1474-1G TCIAC1474-25G TCIAC1474-5G
Ethyl (R)-(-)-4-cyano-3-hydroxybutyate ≥97.0% (by GC)
Ethyl (R)-(-)-4-cyano-3-hydroxybutyate
Formel: C₇H₁₁NO₃
MW: 157,169 g/mol
MDL nummer: MFCD00270839
CAS-nummer: 141942-85-0
EINECS: 430-220-6

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %
Specific rotation [a]20/D -31.0 to -37.0 deg (C=1, CHCL3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR