Chloromethyl phenyl sulphone ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: ((Chloromethyl)sulphonyl)benzene, 1-(Chloromethylsulphonyl)benzene, Phenyl Chloromethyl Sulphone

C1571-25G C1571-5G
TCIAC1571-25GEA 4925 NOK
TCIAC1571-25G TCIAC1571-5G
Chloromethyl phenyl sulphone ≥98.0% (by GC)
Chloromethyl phenyl sulphone
Formel: C₇H₇ClO₂S
MW: 190,64 g/mol
Kokepunkt: 130°C
MDL nummer: MFCD00007551
CAS-nummer: 7205-98-3
EINECS: 230-581-8

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 51.0 to 55.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR