5-Chloro-2-mercaptobenzimidazole ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Danger

Synonymer: 5-Chloro-2-benzimidazolethiol

C1590-25G C1590-5G
TCIAC1590-25GEA 4075 NOK
TCIAC1590-25G TCIAC1590-5G
5-Chloro-2-mercaptobenzimidazole ≥98.0% (by titrimetric analysis)
5-Chloro-2-mercaptobenzimidazole
Formel: C₇H₅ClN₂S
MW: 184,64 g/mol
MDL nummer: MFCD01658762
CAS-nummer: 25369-78-2
EINECS: 246-903-5
FN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ff2e55c213ee3c740e22b411814dc17711b8fd4b&vid=06feeb222956d1b6018358d79f1e1577920a0eb2&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR