N-Carbobenzyloxy-D-proline ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI

C1730-25G C1730-5G
TCIAC1730-25GEA 1700 NOK
TCIAC1730-25G TCIAC1730-5G
N-Carbobenzyloxy-D-proline ≥98.0% (by HPLC, titration analysis)
N-Carbobenzyloxy-D-proline
Formel: C₁₃H₁₅NO₄
MDL nummer: MFCD00063228
CAS-nummer: 6404-31-5
EINECS: 229-021-5

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 74.0 to 78.0 deg-C
Specific rotation [a]20/D +37.0 to +43.0 deg (C=2, EtOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR