4-Chloro-1,3-dioxolan-2-one ≥95.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 4-Chloro-2-oxo-1,3-dioxolane, Chloroethylene Carbonate

C1858-25G C1858-5G
TCIAC1858-25GEA 3075 NOK
TCIAC1858-25G TCIAC1858-5G
4-Chloro-1,3-dioxolan-2-one ≥95.0% (by GC)
4-Chloro-1,3-dioxolan-2-one
Formel: C₃H₃ClO₃
MW: 122,50 g/mol
Kokepunkt: 123°C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00005383
CAS-nummer: 3967-54-2
EINECS: 223-583-5

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Red to Green clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR