(1ɑ,2ɑ,4ɑ)-1,2,4-Cyclohexanetricarboxylic acid ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

C2062-25G C2062-5G
TCIAC2062-25GEA 2950 NOK
TCIAC2062-25G TCIAC2062-5G
(1ɑ,2ɑ,4ɑ)-1,2,4-Cyclohexanetricarboxylic acid ≥98.0% (by GC, titration analysis)
(1alpha,2alpha,4alpha)-1,2,4-Cyclohexanetricarboxylic acid
Formel: C₉H₁₂O₆
MW: 216,19 g/mol
CAS-nummer: 76784-95-7

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 180.0 to 184.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR