(1ɑ,3ɑ,5ɑ)-1,3,5-Cyclohexanetricarboxylic acid ≥98.0% (by titrimetric analysis)
Leverandør: TCI
|
Warning
|
Synonymer:
Cis,cis-cyclohexane-1,3,5-tricarboxylic acid, (1α,3α,5α)-1,3,5-Cyclohexanetricarboxylic acid
C2063-25G
C2063-5G
TCIAC2063-25GEA
3550
NOK
TCIAC2063-25G
TCIAC2063-5G
(1ɑ,3ɑ,5ɑ)-1,3,5-Cyclohexanetricarboxylic acid ≥98.0% (by titrimetric analysis)
(1alpha,3alpha,5alpha)-1,3,5-Cyclohexanetricarboxylic acid
Formel:
C₉H₁₂O₆ MW: 216,19 g/mol |
MDL nummer:
MFCD00075294 CAS-nummer: 16526-68-4 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 96.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 215.0 to 221.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...