2-Cyanoethyl-N,N,N',N'-tetraisopropylphosphordiamidite ≥95.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: Bis(diisopropylamino)(2-cyanoethoxy)phosphine, 3-((Bis(diisopropylamino)phosphino)oxy)propanenitrile

C2228-1G C2228-5G
TCIAC2228-1GEA 475 NOK
TCIAC2228-1G TCIAC2228-5G
2-Cyanoethyl-N,N,N',N'-tetraisopropylphosphordiamidite ≥95.0% (by GC, titration analysis)
2-Cyanoethyl-N,N,N',N'-tetraisopropylphosphordiamidite
Formel: C₁₅H₃₂N₃OP
MW: 301,41 g/mol
Flash punkt: 60°C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00012213
CAS-nummer: 102691-36-1
FN: 1993
ADR: 3,III

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 95.0 %
Purity (Nonaqueous Titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR