Clindamycin phosphate ≥97.0% (by titrimetric analysis)

Leverandør: TCI
Warning

C2257-25G C2257-5G
TCIAC2257-25GEA 2025 NOK
TCIAC2257-25G TCIAC2257-5G
Clindamycin phosphate ≥97.0% (by titrimetric analysis)
Clindamycin phosphate
Formel: C₁₈H₃₄ClN₂O₈PS
Smeltepunkt: 114°C
MDL nummer: MFCD07793328
CAS-nummer: 24729-96-2
EINECS: 246-433-0
Merck Indeks: 14,02356

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 97.0 % (calcd.on anh.substance)
Specific rotation [a]20/D +115.0 to +130.0 deg (C=1, H2O) (calcd.on anh.substance)
Water max. 6.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR