3-Chloropyridine N-Oxide ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: 3-Chloropyridine 1-oxide

C2329-1G C2329-5G
TCIAC2329-1GEA 1075 NOK
TCIAC2329-1G TCIAC2329-5G
3-Chloropyridine N-Oxide ≥98.0% (by GC, titration analysis)
3-Chloropyridine N-Oxide
Formel: C₅H₄ClNO
MW: 129,54 g/mol
Kokepunkt: 89°C
MDL nummer: MFCD00128857
CAS-nummer: 1851-22-5

Bestill nå

Spesifikasjon Testresultater

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 56.0 to 60.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR