Cyromazine ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
|
Warning
|
Synonymer:
2,4-Diamino-6-(cyclopropylamino)-1,3,5-triazine, N2-Cyclopropylmelamine
C2366-25G
C2366-5G
TCIAC2366-25GEA
1875
NOK
TCIAC2366-25G
TCIAC2366-5G
Cyromazine ≥98.0% (by HPLC, titration analysis)
Cyromazine
Formel:
C₆H₁₀N₆ MW: 166,19 g/mol |
MDL nummer:
MFCD00078650 CAS-nummer: 66215-27-8 EINECS: 266-257-8 Merck Indeks: 14,02775 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 222.0 to 226.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...