(-)-Corey lactone benzoate ≥98.0% (by HPLC)

Leverandør: TCI

Synonymer: (3aR,4S,5R,6aS)-4-(Hydroxymethyl)-2-oxohexahydro-2H-cyclopenta[b]furan-5-yl Benzoate, (3aR,4S,5R,6aS)-5-(Benzoyloxy)hexahydro-4-(hydroxymethyl)-2H-cyclopenta[b]furan-2-one

C2605-200MG C2605-1G
TCIAC2605-200MGEA 785 NOK
TCIAC2605-200MG TCIAC2605-1G
(-)-Corey lactone benzoate ≥98.0% (by HPLC)
(-)-Corey lactone benzoate
Formel: C₁₅H₁₆O₅
MW: 276,29 g/mol
MDL nummer: MFCD00674102
CAS-nummer: 39746-00-4
EINECS: 254-615-6

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 118.0 to 122.0 deg-C
Specific rotation [a]20/D -72.0 to -77.0 deg (C=1、CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR