2-Chloro-4,6-dimethylnicotinonitrile ≥98.0% (by GC)
Leverandør: TCI
|
Danger
|
Synonymer:
2-Chloro-3-cyano-4,6-dimethylpyridine, 2-Chloro-4,6-dimethylpyridine-3-carbonitrile
C2668-5G
C2668-25G
TCIAC2668-5GEA
380
NOK
TCIAC2668-5G
TCIAC2668-25G
2-Chloro-4,6-dimethylnicotinonitrile ≥98.0% (by GC)
2-Chloro-4,6-dimethylnicotinonitrile
Formel:
C₈H₇ClN₂ MW: 166,61 g/mol |
MDL nummer:
MFCD00051676 CAS-nummer: 14237-71-9 FN: 3439 ADR: 6.1,III |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 97.0 to 101.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...