1-Chloro-4-ethynylbenzene ≥98.0% (by GC)

Leverandør: TCI
Danger

Synonymer: (4-Chlorophenyl)acetylene, 1-Ethynyl-4-chlorobenzene, (4-Chlorophenyl)ethyne

C2670-5G C2670-1G
TCIAC2670-5GEA 1325 NOK
TCIAC2670-5G TCIAC2670-1G
1-Chloro-4-ethynylbenzene ≥98.0% (by GC)
1-Chloro-4-ethynylbenzene
Formel: C₈H₅Cl
MW: 136,58 g/mol
Kokepunkt: 60°C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00191917
CAS-nummer: 873-73-4
FN: 1325
ADR: 4.1,II

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 45.0 to 49.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR