4,6-Dimethyl-2-oxo-1,2-dihydropyridine-3-carbonitrile ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
C2672-5G
C2672-25G
TCIAC2672-5GEA
220
NOK
TCIAC2672-5G
TCIAC2672-25G
4,6-Dimethyl-2-oxo-1,2-dihydropyridine-3-carbonitrile ≥98.0% (by GC)
4,6-Dimethyl-2-oxo-1,2-dihydropyridine-3-carbonitrile
Formel:
C₈H₈N₂O MW: 148,17 g/mol |
MDL nummer:
MFCD00006269 CAS-nummer: 769-28-8 EINECS: 212-207-5 |
Spesifikasjon Testresultater
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...