4-Amino-ɑ-(tert-butylaminomethyl)-3,5-dichlorobenzyl alcohol hydrochloride ≥97.0% (by HPLC, total nitrogen)

Leverandør: TCI
Danger

Synonymer: Clenbuterol hydrochloride

C2691-100MG C2691-500MG
TCIAC2691-100MGEA 1500 NOK
TCIAC2691-100MG TCIAC2691-500MG
4-Amino-ɑ-(tert-butylaminomethyl)-3,5-dichlorobenzyl alcohol hydrochloride ≥97.0% (by HPLC, total nitrogen)
4-Amino-alpha-(tert-butylaminomethyl)-3,5-dichlorobenzyl alcohol hydrochloride
Formel: C₁₂H₁₈Cl₂N₂O
MW: 313,65 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00083280
CAS-nummer: 21898-19-1
EINECS: 244-643-7
FN: 2811
ADR: 6.1,III
Merck Indeks: 14,02347

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (with Total Nitrogen) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR