(S)-Benzyl(2-oxotetrahydrofuran-3-yl)carbamate ≥98.0% (by HPLC, total nitrogen)

Leverandør: TCI

Synonymer: N-Cbz-L-homoserine Lactone, (S)-alpha-(Carbobenzoxyamino)-gamma-butyrolactone, Cbz-Hse-Lactone, (S)-alpha-(Benzyloxycarbonylamino)-gamma-butyrolactone, N-Benzyloxycarbonyl-L-homoserine Lactone, (S)-Benzyl (2-oxotetrahydrofuran-3-yl)carbamate, N-Carbobenzoxy-L-homoserine Lactone, (S)-alpha-(Cbz-amino)-gamma-butyrolactone

C2699-1G C2699-5G
TCIAC2699-1GEA 460 NOK
TCIAC2699-1G TCIAC2699-5G
(S)-Benzyl(2-oxotetrahydrofuran-3-yl)carbamate ≥98.0% (by HPLC, total nitrogen)
(S)-Benzyl(2-oxotetrahydrofuran-3-yl)carbamate
Formel: C₁₂H₁₃NO₄
MW: 235,2400 g/mol
MDL nummer: MFCD00674555
CAS-nummer: 35677-89-5

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 127.0 to 132.0 deg-C
Specific rotation [a]20/D -30.0 to -34.0 deg (C=1、MeOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR